Order Entry
Netherlands
ContactUsLinkComponent
Anti-CD80 Armenian hamster Monoclonal Antibody [clone: 16-10A1] (APC (Allophycocyanin))
Anti-CD80 Armenian hamster Monoclonal Antibody [clone: 16-10A1] (APC (Allophycocyanin))
Catalog # ABCAAB95549-50
Supplier:  Abcam
undefined
Anti-CD80 Armenian hamster Monoclonal Antibody [clone: 16-10A1] (APC (Allophycocyanin))
Catalog # ABCAAB95549-50
Supplier:  Abcam

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    CD80 molecule
  • Antigen symbol:
    CD80
  • Clonality:
    Monoclonal
  • Clone:
    16-10A1
  • Conjugation:
    APC (Allophycocyanin)
  • Flow cytometry:
    Yes
  • Host:
    Armenian hamster
  • Isotype:
    IgG
  • Reactivity:
    Mouse
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    50 µg
  • Pack type:
    Vial
  • Pk:
    50 µG

Specifications

About this item

APC Armenian hamster monoclonal [16-10A1] to CD80.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: CD80
Clonality: Monoclonal
Clone: 16-10A1
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Armenian hamster
Isotype: IgG
Reactivity: Mouse