Order Entry
Netherlands
ContactUsLinkComponent
Anti-Syntaxin 1a (phospho S14) Rabbit Polyclonal Antibody
Anti-Syntaxin 1a (phospho S14) Rabbit Polyclonal Antibody
  ABCAAB63571-100
 :  Abcam
undefined
Anti-Syntaxin 1a (phospho S14) Rabbit Polyclonal Antibody
  ABCAAB63571-100
 :  Abcam

 

  • Antibody type:
    Primary
  • Antigen name:
    Syntaxin 1A
  • Antigen symbol:
    STX1A
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Seq:
    1286
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µg
  • Pack type:
    Vial
  • Pk:
    100 µG

 

 

Rabbit polyclonal to Syntaxin 1a (phospho S14).

 :

All products are FOR RESEARCH USE ONLY.   PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES.  Customer is granted a limited use non-exclusive licence for its own internal research.  Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product.  By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: Syntaxin 1a
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse