Anti-HSP110 Rabbit Polyclonal Antibody
Catalogus # BIRBORB95896-50
Supplier: Biorbyt
Specifications
- Antibody type:Primary
- Antigen symbol:HSP110
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Western blot:Yes
- Storage buffer:PBS, pH 7,2, 50% glycerol, 0,09% sodium azide
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by affinity chromatography
- Size:50 µg
- Pk:50 µG
Specifications
About this item
Immunochemistry: 5 ?g/ml; Western Blot: 1:1000
Type: Primary
Antigen: HSP110
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: