Anti-CASK Rabbit Polyclonal Antibody
Catalog # BIRBORB75814-100
Supplier: Biorbyt
Specifications
- Antibody type:Primary
- Antigen name:calcium/calmodulin-dependent serine Protein kinase (MAGUK family)
- Antigen symbol:CASK
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Storage temperature:Cool
- Concentration:1 mg/mL
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: CASK (calcium/calmodulin-dependent serine protein kinase (MAGUK family))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse, Rat