Anti-MMP2 Rabbit Polyclonal Antibody (RBITC)
BIRBORB131514-100
: Biorbyt
- Antibody type:Primary
- Antigen name:matrix metallopeptidase 2 (gelatinase A, 72kDa gelatinase, 72kDa type IV collagenase)
- Antigen symbol:MMP2
- Clonality:Polyclonal
- Conjugation:RBITC (Rhodamine B Isothiocyanate)
- Host:Rabbit
- Isotype:IgG
- Form:liquid
- Storage buffer:10 mM PBS, pH 7,4 with 10 mg/ml BSA, 0,03% Proclin 300 and 25% glycerol
- Molecular weight:72 kDa
- Storage temperature:Cool
- Shipping temperature:Ice
- Immunogen:Epitope mapping near the N-terminus of MMP-2 of human, mouse and rat
- Purification:purified by affinity chromatography
- Size:100 µl
- Pk:100 µl
Type: Primary
Antigen: MMP2 (matrix metallopeptidase 2 (gelatinase A, 72kDa gelatinase, 72kDa type IV collagenase))
Clonality: polyclonal
Clone:
Conjugation: RBITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: