Anti-PKM Rabbit Polyclonal Antibody (PE (Phycoerythrin)/Cy3®)
BIRBORB129341-100
: Biorbyt
- Antibody type:Primary
- Antigen name:pyruvate kinase, muscle
- Antigen symbol:PKM
- Clonality:Polyclonal
- Conjugation:PE (Phycoerythrin)/Cy3®
- Host:Rabbit
- Isotype:IgG
- Reactivity:
- Environmentally Preferable:
- Form:liquid
- Modification:phospho-Tyr105
- Storage buffer:10 mM PBS, pH 7,4 with 10 mg/ml BSA, 0,03% Proclin 300 and 25% glycerol
- Molecular weight:58 kDa
- Storage temperature:Cool
- Shipping temperature:Ice
- Immunogen:KLH conjugated synthetic phosphopeptide derived from human PKM2 around the phosphorylation site of Tyr105.
- Purification:purified by affinity chromatography
- Size:100 µl
- Pk:100 µl
Type: Primary
Antigen: PKM (pyruvate kinase, muscle)
Clonality: polyclonal
Clone:
Conjugation: PE (Phycoerythrin)/Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: