Anti-RAR Beta Rabbit Polyclonal Antibody
BIRBORB109224-50
: Biorbyt
- Antibody type:Primary
- Antigen name:Retinoic Acid Receptor Beta
- Antigen symbol:RARB
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Storage temperature:Cool
- Shipping temperature:Ice
- Immunogen:C-terminal region of human
- Purification:purified by immunoaffinity chromatography
- Size:50 µg
- Pk:50 µG
Type: Primary
Antigen: RAR Beta
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human