Anti-MMP2 Rabbit Polyclonal Antibody (FITC (Fluorescein))
BIRBORB103021-100
: Biorbyt
- Type antilichaam:Primair
- antigeen symbool:MMP2
- Clonality:Polyklonaal
- Conjugatie:FITC (Fluoresceïne-isothiocyanaat)
- Host:Rabbit
- ImmunoFluorescentie:Yes
- Isotype:IgG
- Formulier:liquid
- Opslagbuffer:10 mM PBS, pH 7,4 with 10 mg/ml BSA, 0,03% Proclin 300 and 25% glycerol
- Moleculair gewicht:72 kDa
- Opslagtemperatuur:Cool
- Concentratie:0,5 mg/ml
- Shipping temperature:Ice
- Purification:purified by affinity chromatography
- Maat:100 µg
- VPE:100 µG
Immunofluorescence: 1:50-200 (based on 0.5 mg/ml)
Type: Primary
Antigen: MMP2 (matrix metallopeptidase 2 (gelatinase A, 72kDa gelatinase, 72kDa type IV collagenase))
Clonality: polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: