Anti-COL4A2 Rabbit Polyclonal Antibody
BIRBORB47988-100
: Biorbyt
- Antibody type:Primary
- Antigen name:collagen, type IV, alpha 2
- Antigen symbol:COL4A2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:lyophilized
- Amino acid number:1697 to 1712
- Storage buffer:5mg BSA, 0,9mg NaCl, 0,2mg Na2HPO4, 0,05mg Thimerosal, 0,05mg NaN3.
- Sequence:LIRTHISRCQVCMKNL
- Storage temperature:Cool
- Concentration:500 ug/ml
- Shipping temperature:Ice
- Immunogen:A synthetic peptide corresponding to a sequence at the C-terminus of human COL4A2(1697-1712aa LIRTHISRCQVCMKNL), identical to the related rat and mouse sequences.
- Purification:purified by immunoaffinity chromatography
- Size:100 µg
- Pk:100 µG
Type: Primary
Antigen: COL4A2 (collagen, type IV, alpha 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity: Human, Mouse, Rat