Order Entry
Netherlands
ContactUsLinkComponent
Anti-RPA 14 kDa subunit Rabbit Polyclonal Antibody
Anti-RPA 14 kDa subunit Rabbit Polyclonal Antibody
Catalog # BIRBORB40745-100
Supplier:  Biorbyt
Anti-RPA 14 kDa subunit Rabbit Polyclonal Antibody
Catalog # BIRBORB40745-100
Supplier:  Biorbyt

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    RPA 14 kDa subunit
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Amino acid number:
    1 to 91
  • Storage buffer:
    1XPBS, 1%BSA, 20% Glycerol (pH7).
  • Storage temperature:
    Cool
  • Concentration:
    1 mg/ml
  • Shipping temperature:
    Ice
  • Immunogen:
    Recombinant protein fragment contain a sequence corresponding to a region within amino acids 1 and 91 of RPA 14 kDa subunit .
  • Purification:
    purified by antigen affinity chromatography
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Western Blot: 1:500-1:3000; Immunochemistry: 1:100-1:250

Type: Primary
Antigen: RPA 14 kDa subunit
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human