Anti-IgA/IgD/IgE/IgG/IgM Rabbit Polyclonal Antibody
BIRBORB22360-1
: Biorbyt
- Antibody type:Secondary
- Antigen symbol:IgA/IgD/IgE/IgG/IgM
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:
- Reactivity:
- Environmentally Preferable:
- Format:antiserum
- Form:lyophilized
- Storage temperature:Cool
- Shipping temperature:Ice
- Immunogen:Purified normal Ig fractions isolated from pooled human serum. Freunds complete adjuvant is used in the first step of the immunization procedure.
- Size:1 ml
- Pk:1 mL
Type: Secondary
Antigen: IgG/IgA/IgM/IgD/IgE
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: