Anti- IgG Antibody (Unconjugated)
Catalog # BIRBORB22143-100
Supplier: Biorbyt
Specifications
- Antibody type:Secondary
- Antigen symbol:IgG
- Conjugation:Unconjugated
- Host:
- Isotype:
- Reactivity:
- Environmentally Preferable:
- Form:lyophilized
- Storage buffer:PBS, pH 7,2
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 mg
- Pk:100 mg
Specifications
About this item
Type: Secondary
Antigen: IgG (Goat) (goat IgG)
Clonality:
Clone:
Conjugation: unconjugated
Epitope:
Host: None
Isotype: none
Reactivity: