Anti-IgA/IgG/IgM Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
BIRBORB21534-1
: Biorbyt
- Antibody type:Secondary
- Antigen symbol:IgA/IgG/IgM
- Clonality:Polyclonal
- Conjugation:HRP (Horseradish Peroxidase)
- Host:Rabbit
- Isotype:
- Reactivity:
- Environmentally Preferable:
- Format:immunoconjugate
- Form:lyophilized
- Storage temperature:Cool
- Concentration:1 mg/ml, Enzyme/IgG protein molar ratio (E/P) is approximately 1,7, no foreign proteins added,
- Shipping temperature:Ice
- Immunogen:Purified immunoglobulin IgG, IgA and IgM fractions isolated from pooled rhesus monkey serum. Freunds complete adjuvant is used in the first step of the immunization procedure.
- Size:1 ml
- Pk:1 mL
Type: Secondary
Antigen: Monkey IgG IgA IgM
Clonality: polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: none
Reactivity: