Anti-IgG Rabbit Polyclonal Antibody (TRITC (Tetramethylrhodamine Isothiocyanate))
Catalog # BIRBORB21500-2
Supplier: Biorbyt
Specifications
- Antibody type:Secondary
- Antigen symbol:IgG
- Clonality:Polyclonal
- Conjugation:TRITC (Tetramethylrhodamine Isothiocyanate)
- Host:Rabbit
- Isotype:
- Reactivity:
- Environmentally Preferable:
- Format:antiserum
- Form:lyophilized
- Storage temperature:Cool
- Concentration:1 mg/ml, Fluorochrome/IgG protein molar ratio (F/P) is approximately 1,8, no foreign proteins added,
- Shipping temperature:Ice
- Immunogen:Purified normal IgG isolated from pooled goat serum. Freunds complete adjuvant is used in the first step of the immunization procedure.
- Size:2 ml
- Pk:2 mL
Specifications
About this item
Type: Secondary
Antigen: IgG (Goat) (goat IgG)
Clonality: polyclonal
Clone:
Conjugation: TRITC (Rhodamine)
Epitope:
Host: Rabbit
Isotype: none
Reactivity: