Anti-alpha Chymotrypsinogen A Rabbit Polyclonal Antibody (Biotin)
BIRBORB20930-1
: Biorbyt
- Antibody type:Primary
- Antigen name:alpha Chymotrypsinogen A
- Clonality:Polyclonal
- Conjugation:Biotin
- Host:Rabbit
- Isotype:
- Reactivity:Bovine
- Environmentally Preferable:
- Format:immunoconjugate
- Storage buffer:phosphate buffered saline
- Storage temperature:Cool
- Concentration:1 mg/ml, Biotin/IgG protein molar ratio (B/P) approximately 6,5, no foreign proteins added,
- Shipping temperature:Ice
- Immunogen:alpha Chymotrypsinogen A isolated and purified from bovine pancreas. Freunds complete adjuvant is used in the first step of the immunization procedure.
- Size:1 ml
- Pk:1 mL
Type: Primary
Antigen: alpha Chymotrypsinogen A
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Bovine