Anti-Integrin avb3 Rabbit Polyclonal Antibody (FITC (Fluorescein))
Catalog # BIRBORB15865-100
Supplier: Biorbyt
Specifications
- Type antilichaam:Primair
- Clonality:Polyklonaal
- Conjugatie:FITC (Fluoresceïne-isothiocyanaat)
- Host:Rabbit
- ImmunoFluorescentie:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Formulier:liquid
- Opslagbuffer:10 mM PBS, pH 7,4 with 10 mg/ml BSA, 0,03% Proclin 300 and 25% glycerol
- Opslagtemperatuur:Cool
- Concentratie:0,5 mg/ml
- Shipping temperature:Ice
- Immunogeen:Synthetic peptide
- Purification:purified by affinity chromatography
- Maat:100 µg
- VPE:100 µG
Specifications
About this item
Immunofluorescence: 1:50-200 (based on 0.5 mg/ml)
Type: Primary
Antigen: Integrin avb3
Clonality: polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat