Order Entry
Netherlands
ContactUsLinkComponent
 
Anti-AOC3 Rabbit Polyclonal Antibody
 :  Biorbyt
 
Anti-AOC3 Rabbit Polyclonal Antibody
 :  Biorbyt

 

  • Catalog No:
  • BIRBORB11547-200
  • BIRBORB11547-100
  • Antigen symbol:
  • AOC3
  • AOC3
  • Antigen name:
  • amine oxidase, copper containing 3
  • amine oxidase, copper containing 3
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Polyclonal
  • Polyclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Reactivity:
  • Human, Rat, Mouse
  • Human, Rat, Mouse
  • Host:
  • Rabbit
  • Rabbit
  • Isotype:
  • IgG
  • IgG
  • Western blot:
  • Yes
  • Yes
  • ELISA:
  • Yes
  • Yes
  • ImmunoChemistry:
  • Yes
  • Yes
  • Storage temperature:
  • Cool
  • Cool
  • Shipping temperature:
  • Ice
  • Ice

 

Frequently Bought Together