Bestelling invoeren
Netherlands
ContactUsLinkComponent
Anti-MECP2 Rabbit Polyclonal Antibody (FITC (Fluorescein))
Anti-MECP2 Rabbit Polyclonal Antibody (FITC (Fluorescein))
Catalog # BIRBORB9255-100
Supplier:  Biorbyt
Anti-MECP2 Rabbit Polyclonal Antibody (FITC (Fluorescein))
Catalog # BIRBORB9255-100
Supplier:  Biorbyt

Specifications

  • Type antilichaam:
    Primair
  • antigeen symbool:
    MECP2
  • Clonality:
    Polyklonaal
  • Conjugatie:
    FITC (Fluoresceïne-isothiocyanaat)
  • Host:
    Rabbit
  • ImmunoFluorescentie:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Formulier:
    liquid
  • Opslagbuffer:
    10 mM PBS, pH 7,4 with 10 mg/ml BSA, 0,03% Proclin 300 and 25% glycerol
  • Moleculair gewicht:
    55 kDa
  • Opslagtemperatuur:
    Cool
  • Concentratie:
    0,5 mg/ml
  • Shipping temperature:
    Ice
  • Immunogeen:
    KLH conjugated synthetic peptide derived from human MeCP2.
  • Purification:
    purified by affinity chromatography
  • Maat:
    100 µg
  • VPE:
    100 µG

Specifications

About this item

Immunofluorescence: 1:50-200 (based on 0.5 mg/ml)

Type: Primary
Antigen: MECP2 (methyl CpG binding protein 2 (Rett syndrome))
Clonality: polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat