Order Entry
Netherlands
ContactUsLinkComponent
 
Anti-NEDD1 Mouse Monoclonal Antibody [clone: 7D10]
  ABNOH00121441-M05
 :  Abnova
undefined
Anti-NEDD1 Mouse Monoclonal Antibody [clone: 7D10]
  ABNOH00121441-M05
 :  Abnova

 

  • Antibody type:
    Primary
  • Antigen name:
    Neural Precursor Cell Expressed, Developmentally Down-Regulated 1
  • Antigen symbol:
    NEDD1
  • Clonality:
    Monoclonal
  • Clone:
    7D10
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • Isotype:
  • Reactivity:
    Human,
    Mouse
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Cross adsorption:
    No
  • Gene ID:
    121441
  • Antigen synonyms:
    FLJ35902|developmentally down regulated gene 1|Neural precursor cell expressed|GCP WD|NEDD1_HUMAN.|TUBGCP7
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µg
  • Pk:
    100 µg

 

 

Type: Primary
Antigen: NEDD1
Clonality: Monoclonal
Clone: 7D10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human, Mouse