Order Entry
Netherlands
ContactUsLinkComponent
Anti-DDX54 Mouse Monoclonal Antibody [clone: 2E4]
Anti-DDX54 Mouse Monoclonal Antibody [clone: 2E4]
Catalog # ABNOH00079039-M02
Supplier:  Abnova
undefined
Anti-DDX54 Mouse Monoclonal Antibody [clone: 2E4]
Catalog # ABNOH00079039-M02
Supplier:  Abnova

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    DEAD (Asp-Glu-Ala-Asp) box polypeptide 54
  • Antigen symbol:
    DDX54
  • Clonality:
    Monoclonal
  • Clone:
    2E4
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Gene ID:
    79039
  • Antigen synonyms:
    DP97
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    50 µg
  • Pk:
    50 µg

Specifications

About this item

Type: Primary
Antigen: DDX54
Clonality: Monoclonal
Clone: 2E4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human