Order Entry
Netherlands
ContactUsLinkComponent
Anti-FAM64A Rabbit Polyclonal Antibody
Anti-FAM64A Rabbit Polyclonal Antibody
Catalog # ABNOH00054478-D02P
Supplier:  Abnova
undefined
Anti-FAM64A Rabbit Polyclonal Antibody
Catalog # ABNOH00054478-D02P
Supplier:  Abnova

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Family with sequence similarity 64, member A
  • Antigen symbol:
    FAM64A
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Gene ID:
    54478
  • Antigen synonyms:
    RCS1|CATS
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µg
  • Pk:
    100 µg

Specifications

About this item

Type: Primary
Antigen: FAM64A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human