Anti-IKZF2 Mouse Polyclonal Antibody
Catalogus # ABNOH00022807-B01P
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Ikaros family Zinc Finger Protein 2
- Antigen symbol:IKZF2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoFluorescence:Yes
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:22807
- Antigen synonyms:HELIOS|ZNF1A2|ANF1A2|ZNFN1A2|Ikaros family zinc finger 2 (Helios)
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:50 µg
- Pk:50 µg
Specifications
About this item
Type: Primary
Antigen: IKZF2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human