Anti-RPP14 Mouse Monoclonal Antibody [clone: 3H4]
Catalogus # ABNOH00011102-M01A
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Ribonuclease P/MRP 14kDa Subunit
- Antigen symbol:RPP14
- Clonality:Monoclonal
- Clone:3H4
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:11102
- Antigen synonyms:P14
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:200 µl
- Pk:200 µl
Specifications
About this item
Type: Primary
Antigen: RPP14
Clonality: Monoclonal
Clone: 3H4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human