Anti-CCT6B Mouse Monoclonal Antibody [clone: 1A4]
Catalog # ABNOH00010693-M01
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Chaperonin Containing TCP1, subunit 6B (zeta 2)
- Antigen symbol:CCT6B
- Clonality:Monoclonal
- Clone:1A4
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:10693
- Antigen synonyms:CCT-zeta-2|CCTZ-2|TSA303|Cctz2|TCP-1-zeta-2
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: CCT6B
Clonality: Monoclonal
Clone: 1A4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human