Anti-CLDN16 Mouse Monoclonal Antibody [clone: 1F2]
Catalogus # ABNOH00010686-M02
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Claudin 16
- Antigen symbol:CLDN16
- Clonality:Monoclonal
- Clone:1F2
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:10686
- Antigen synonyms:HOMG3|PCLN1
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: CLDN16
Clonality: Monoclonal
Clone: 1F2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human