Anti-AKAP8 Mouse Monoclonal Antibody [clone: 3D4]
ABNOH00010270-M01
: Abnova
- Antibody type:Primary
- Antigen name:A-kinase anchor protein 8
- Antigen symbol:AKAP8
- Clonality:Monoclonal
- Clone:3D4
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:10270
- Antigen synonyms:AKAP95|AKAP-8|AKAP-95|AKAP 95|AKAP 8|A-kinase anchor protein 95 kDa
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Type: Primary
Antigen: AKAP8
Clonality: Monoclonal
Clone: 3D4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human