Order Entry
Netherlands
ContactUsLinkComponent
Anti-AOC3 Mouse Monoclonal Antibody [clone: 4B8]
Anti-AOC3 Mouse Monoclonal Antibody [clone: 4B8]
  ABNOH00008639-M06
 :  Abnova
undefined
Anti-AOC3 Mouse Monoclonal Antibody [clone: 4B8]
  ABNOH00008639-M06
 :  Abnova

 

  • Antibody type:
    Primary
  • Antigen name:
    Amine Oxidase, Copper Containing 3
  • Antigen symbol:
    AOC3
  • Clonality:
    Monoclonal
  • Clone:
    4B8
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • Reactivity:
    Human
  • Cross adsorption:
    No
  • Gene ID:
    8639
  • Antigen synonyms:
    VAP-1.|Vascular adhesion protein 1|Semicarbazide-sensitive amine oxidase|Membrane primary amine oxidase|Membrane copper amine oxidase|Semicarbazide sensitive amine oxidase|Vascular Adhesion Protein 1|HPAO|SSAO|AOC3|VP97|AOC3_HUMAN|Amine Oxidase|Copper Containing 3 (vascular adhesion protein 1)|Copper amine oxidase
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µg
  • Pk:
    100 µg

 

 

Type: Primary
Antigen: AOC3
Clonality: Monoclonal
Clone: 4B8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human