Anti-AOC3 Mouse Polyclonal Antibody
Catalog # ABNOH00008639-B01P
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Amine Oxidase, Copper Containing 3
- Antigen symbol:AOC3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:8639
- Antigen synonyms:VAP-1.|Vascular adhesion protein 1|Semicarbazide-sensitive amine oxidase|Membrane primary amine oxidase|Membrane copper amine oxidase|Semicarbazide sensitive amine oxidase|Vascular Adhesion Protein 1|HPAO|SSAO|AOC3|VP97|AOC3_HUMAN|Amine Oxidase|Copper Containing 3 (vascular adhesion protein 1)|Copper amine oxidase
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:50 µg
- Pk:50 µg
Specifications
About this item
Type: Primary
Antigen: AOC3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human