Anti-IFITM1 Mouse Polyclonal Antibody
ABNOH00008519-B02
: Abnova
- Antibody type:Primary
- Antigen name:Interferon induced transmembrane protein 1
- Antigen symbol:IFITM1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:8519
- Antigen synonyms:Interferon-induced transmembrane protein 1|IFITM1|DSPA2a|Dispanin subfamily A member 2a|IFI17|9-27|CD225|LEU13
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:50 µl
- Pk:50 µl
Type: Primary
Antigen: IFITM1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human