Anti-TCL1A Mouse Monoclonal Antibody [clone: 1E8]
Catalog # ABNOH00008115-M01
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:T-cell leukemia/lymphoma 1A
- Antigen symbol:TCL1A
- Clonality:Monoclonal
- Clone:1E8
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Cross adsorption:No
- Gene ID:8115
- Antigen synonyms:TCL1
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: TCL1A
Clonality: Monoclonal
Clone: 1E8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human