Anti-CSDE1 Mouse Polyclonal Antibody
Catalogus # ABNOH00007812-A01
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Cold Shock Domain Containing E1
- Antigen symbol:CSDE1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:7812
- Antigen synonyms:BC016898|AA960392|mKIAA0885|unr|D3Jfr1
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:50 µl
- Pk:50 µl
Specifications
About this item
Type: Primary
Antigen: CSDE1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human