Anti-HNF1B Mouse Monoclonal Antibody [clone: 4E9]
Catalog # ABNOH00006928-M06
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:HNF1 homeobox B
- Antigen symbol:HNF1B
- Clonality:Monoclonal
- Clone:4E9
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Cross adsorption:No
- Gene ID:6928
- Antigen synonyms:HNF1beta|LFB3|HPC11|LF-B3|FJHN|MODY5|HNF-1B|TCF2|TCF-2|HNF-1-beta|VHNF1|HNF2
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: HNF1B
Clonality: Monoclonal
Clone: 4E9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human, Mouse, Rat