Anti-TBX6 Mouse Monoclonal Antibody [clone: 3D6]
Catalogus # ABNOH00006911-M02
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:T-box 6
- Antigen symbol:TBX6
- Clonality:Monoclonal
- Clone:3D6
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Cross adsorption:No
- Gene ID:6911
- Antigen synonyms:TBX6|SCDO5|T-box transcription factor TBX6|T-box protein 6
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: TBX6
Clonality: Monoclonal
Clone: 3D6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human