Anti-MECP2 Mouse Monoclonal Antibody [clone: 2E8]
ABNOH00004204-M04A
: Abnova
- Antibody type:Primary
- Antigen name:Methyl CpG binding protein 2
- Antigen symbol:MECP2
- Clonality:Monoclonal
- Clone:2E8
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:4204
- Antigen synonyms:RTS|MRX79|AUTSX3|MRX16|PPMX|RS|MRXS13|MRXSL|RTT
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:200 µl
- Pk:200 µl
Type: Primary
Antigen: MECP2
Clonality: Monoclonal
Clone: 2E8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human