Anti-MBD1 Mouse Monoclonal Antibody [clone: 4F6]
Catalog # ABNOH00004152-M03
Supplier: Abnova
Specifications
- Type antilichaam:Primair
- antigeen symbool:MBD1
- Clonality:Monoklonaal
- Clone:4F6
- Conjugatie:Onconjugeerd
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Western blot:Yes
- Kruisadsorptie:No
- Gen-ID:4152
- Antigeen synoniemen:CXXC3|PCM1|RFT
- Opslagtemperatuur:Cool
- Shipping temperature:Ice
- Maat:100 µg
- VPE:100 µg
Specifications
About this item
Type: Primary
Antigen: MBD1
Clonality: Monoclonal
Clone: 4F6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human