Anti-SMAD7 Mouse Monoclonal Antibody [clone: 3A12]
Catalog # ABNOH00004092-M11
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:SMAD family member 7
- Antigen symbol:SMAD7
- Clonality:Monoclonal
- Clone:3A12
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:4092
- Antigen synonyms:MADH7|SMAD7|Smad7|MADH8|SMAD family member 7|hSMAD7
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: SMAD7
Clonality: Monoclonal
Clone: 3A12
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human