Anti-GTF2B Rabbit Polyclonal Antibody
Catalog # ABNOH00002959-D01P
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:General transcription factor 2B
- Antigen symbol:GTF2B
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human,Mouse
- Western blot:Yes
- Cross adsorption:No
- Gene ID:2959
- Antigen synonyms:TFIIB|TF2B
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: GTF2B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse