Anti-CTBS Mouse Monoclonal Antibody [clone: 1B5-1B9]
Catalog # ABNOH00001486-M01
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:chitobiase, di-N-acetyl-
- Antigen symbol:CTBS
- Clonality:Monoclonal
- Clone:1B5-1B9
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:1486
- Antigen synonyms:CTB
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: CTBS
Clonality: Monoclonal
Clone: 1B5-1B9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human