Order Entry
Netherlands
ContactUsLinkComponent
Anti-COX4I1 Mouse Monoclonal Antibody [clone: 4A10]
Anti-COX4I1 Mouse Monoclonal Antibody [clone: 4A10]
Catalog # ABNOH00001327-M08
Supplier:  Abnova
undefined
Anti-COX4I1 Mouse Monoclonal Antibody [clone: 4A10]
Catalog # ABNOH00001327-M08
Supplier:  Abnova

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Cytochrome C Oxidase Subunit Iv Isoform 1
  • Antigen symbol:
    COX4I1
  • Clonality:
    Monoclonal
  • Clone:
    4A10
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Gene ID:
    1327
  • Antigen synonyms:
    COXIV|IV-1|AL024441|COX|COX IV-1|Cox4a|Cox4
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µg
  • Pk:
    100 µg

Specifications

About this item

Type: Primary
Antigen: COX4I1
Clonality: Monoclonal
Clone: 4A10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human