Anti-COL4A6 Mouse Monoclonal Antibody [clone: 2F1]
ABNOH00001288-M02
: Abnova
- Antibody type:Primary
- Antigen name:Collagen, type IV, alpha 6
- Antigen symbol:COL4A6
- Clonality:Monoclonal
- Clone:2F1
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Cross adsorption:No
- Gene ID:1288
- Antigen synonyms:CXDELq22.3|DELXq22.3|DFNX6
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Type: Primary
Antigen: COL4A6
Clonality: Monoclonal
Clone: 2F1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human