Anti-COL2A1 Mouse Monoclonal Antibody [clone: 3H1-F9]
ABNOH00001280-M01
: Abnova
- Antibody type:Primary
- Antigen name:Collagen, type II, alpha 1
- Antigen symbol:COL2A1
- Clonality:Monoclonal
- Clone:3H1-F9
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:1280
- Antigen synonyms:SEDC|AOM|STL1|ANFH|COL11A3
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Type: Primary
Antigen: COL2A1
Clonality: Monoclonal
Clone: 3H1-F9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human