Anti-CLCN6 Mouse Monoclonal Antibody [clone: 2H2]
Catalog # ABNOH00001185-M05
Leverancier: Abnova
Specificaties
- Antibody type:Primary
- Antigen name:Chloride Channel, Voltage-sensitive 6
- Antigen symbol:CLCN6
- Clonality:Monoclonal
- Clone:2H2
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:1185
- Antigen synonyms:CLC-6
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specificaties
Over dit artikel
Type: Primary
Antigen: CLCN6
Clonality: Monoclonal
Clone: 2H2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human