Anti-AOC2 Rabbit Polyclonal Antibody
ABNOH00000314-D01P
: Abnova
- Antibody type:Primary
- Antigen name:Amine Oxidase, Copper Containing 2
- Antigen symbol:AOC2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:314
- Antigen synonyms:copper containing 2 retina-specific|RAO|DAO2|amine oxidase
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Type: Primary
Antigen: AOC2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human