Anti-AOC2 Mouse Polyclonal Antibody
Catalog # ABNOH00000314-A01
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Amine Oxidase, Copper Containing 2
- Antigen symbol:AOC2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:314
- Antigen synonyms:copper containing 2 retina-specific|RAO|DAO2|amine oxidase
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:50 µl
- Pk:50 µl
Specifications
About this item
Type: Primary
Antigen: AOC2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human