Order Entry
Netherlands
ContactUsLinkComponent
Anti-ALOX15 Mouse Monoclonal Antibody [clone: 3D8]
Anti-ALOX15 Mouse Monoclonal Antibody [clone: 3D8]
Catalog # ABNOH00000246-M04
Supplier:  Abnova
undefined
Anti-ALOX15 Mouse Monoclonal Antibody [clone: 3D8]
Catalog # ABNOH00000246-M04
Supplier:  Abnova

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Arachidonate 15-lipoxygenase
  • Antigen symbol:
    ALOX15
  • Clonality:
    Monoclonal
  • Clone:
    3D8
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • Isotype:
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Cross adsorption:
    No
  • Gene ID:
    246
  • Antigen synonyms:
    LOG15|Arachidonate 15-lipoxygenase|Arachidonate omega-6 lipoxygenase|15-LOX|ALOX15
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µg
  • Pk:
    100 µg

Specifications

About this item

Type: Primary
Antigen: ALOX15
Clonality: Monoclonal
Clone: 3D8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human