Anti-CDKN2A Mouse Monoclonal Antibody [clone: OTI1F6]
Catalog # ORIGTA802246
Supplier: OriGene
Specifications
- Antibody type:Primary
- Antigen name:Cyclin-dependent kinase inhibitor 2A
- Antigen symbol:CDKN2A
- Clonality:Monoclonal
- Clone:OTI1F6
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:none
- Form:Liquid
- Amino acid number:25 to 156
- Storage buffer:PBS (PH 7.3) containing 1% BSA, 50% glycerol and 0,02% sodium azide.
- Molecular weight:17.8 kDa
- Storage temperature:Cool
- Shipping temperature:Ice
- Immunogen:Protein
- Purification:purified by affinity chromatography
- Pk:100 µl
Specifications
About this item
Recommended Dilutions: Western Blot: 1:500; Immunochemistry: 1:150
Type: Primary
Antigen: CDKN2A (cyclin-dependent kinase inhibitor 2A)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human