Anti-PADI4 Mouse Monoclonal Antibody [clone: OTI6A2]
Catalog # ORIGTA504887
Supplier: OriGene
Specifications
- Antibody type:Primary
- Antigen name:peptidyl arginine deiminase, type IV
- Antigen symbol:PADI4
- Clonality:Monoclonal
- Clone:OTI6A2
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:none
- Form:Liquid
- Amino acid number:299 to 588
- Storage buffer:PBS (PH 7.3) containing 1% BSA, 50% glycerol and 0,02% sodium azide.
- Molecular weight:73.9 kDa
- Storage temperature:Cool
- Shipping temperature:Ice
- Immunogen:Protein
- Purification:purified by affinity chromatography
- Pk:100 µl
Specifications
About this item
Recommended Dilutions: Western Blot: 1:500-1000; Immunochemistry: 1:150; Immunofluorescence: 1:100; Flow Cytometry: 1:100
Type: Primary
Antigen: PADI4 (peptidyl arginine deiminase, type IV)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human