Anti-TRPC5 Mouse Monoclonal Antibody [clone: S67-15]
Catalog # ORIGTA309488
Supplier: OriGene
Specifications
- Antibody type:Primary
- Antigen name:transient receptor potential cation channel, subfamily C, member 5
- Antigen symbol:TRPC5
- Clonality:Monoclonal
- Clone:S67-15
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgG2b
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Cross adsorption:none
- Form:Liquid
- Amino acid number:827 to 845
- Storage buffer:PBS, 0,01% sodium azide
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:100 µG
Specifications
About this item
Recommended Dilutions:
Type: Primary
Antigen: TRPC5 (transient receptor potential cation channel, subfamily C, member 5)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human, Mouse, Rat