Anti-SLC5A5 Mouse Monoclonal Antibody [clone: FP5]
ORIGTA309483
: OriGene
- Antibody type:Primary
- Antigen name:solute carrier family 5 (sodium/iodide cotransporter), member 5
- Antigen symbol:Solute carrier family 5, member 5
- Clonality:Monoclonal
- Clone:FP5
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Human,Rat,Mouse
- Cross adsorption:none
- Form:Liquid
- Amino acid number:468 to 643
- Storage buffer:PBS, 50% glycerol, 0,09% sodium azide
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:100 µG
Recommended Dilutions:
Type: Primary
Antigen: SLC5A5 (solute carrier family 5 (sodium/iodide cotransporter), member 5)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human, Mouse, Rat