Anti-TAAR5 Rabbit Polyclonal Antibody
Catalog # ORIGTA308699
Supplier: OriGene
Specifications
- Antibody type:Primary
- Antigen name:Trace amine associated receptor 5
- Antigen symbol:TAAR5
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:none
- Form:Liquid
- Amino acid number:273 to 337
- Storage buffer:0,1M Tris, 0,1M Glycine, 10% Glycerol (pH7). 0,01% Thimerosal was added as a preservative.
- Molecular weight:38 kDa
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by antigen affinity chromatography
- Pk:100 µl
Specifications
About this item
Recommended Dilutions: Immunochemistry/Immunofluorescence:1:100-1:1000; Western Blot:1:500-1:3000
Type: Primary
Antigen: TAAR5 (trace amine associated receptor 5)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human